Learn More
ABclonal Technology ATP6V0C Rabbit mAb
SDP
Catalog No. 5000006107
Encompass_Preferred
This gene encodes a component of vacuolar ATPase (V-ATPase) a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting zymogen activation receptor-mediated endocytosis and synaptic vesicle proton gradient generation V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain The V1 domain consists of three A and three B subunits two G subunits plus the C D E F and H subunits The V1 domain contains the ATP catalytic site The V0 domain consists of five different subunits a c c -apos c -quot and d This gene encodes the V0 subunit c Alternative splicing results in transcript variants Pseudogenes have been identified on chromosomes 6 and 17
Catalog No. 50-000-06107 Supplier ABclonal Technology Supplier No. A23757100UL
This gene encodes a component of vacuolar ATPase (V-ATPase) a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting zymogen activation receptor-mediated endocytosis and synaptic vesicle proton gradient generation V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain The V1 domain consists of three A and three B subunits two G subunits plus the C D E F and H subunits The V1 domain contains the ATP catalytic site The V0 domain consists of five different subunits a c c -apos c -quot and d This gene encodes the V0 subunit c Alternative splicing results in transcript variants Pseudogenes have been identified on chromosomes 6 and 17