Learn More
Aviva Systems Biology Corp ATP6V0E2 Antibody - middle region (ARP51912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V0E2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP6V0E2. Alternate names: ATP6V0E2L, C7orf32ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60644