Learn More
Aviva Systems Biology Corp ATP6V1B1 Antibody - middle region (ARP56327_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56327_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V1B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP6V1B1. Alternate names: ATP6B1, MGC32642, RTA1B, VATB, VMA2, VPP3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61811