Learn More
Aviva Systems Biology Corp ATP6V1B2 Antibody - N-terminal region (ARP56328_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56328_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V1B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP6V1B2. Alternate names: ATP6B1B2, ATP6B2, HO57, VATB, VPP3, Vma2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61812