Learn More
Aviva Systems Biology Corp ATP6V1C2 Antibody - C-terminal region (ARP62325_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62325_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V1C2. It was validated on Western Blot. . Alternate names: ATP6C2, VMA5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-651