Learn More
Aviva Systems Biology Corp ATP6V1E2 Antibody - middle region (ARP58590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V1E2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ATP6V1E2. Alternate names: ATP6E1, ATP6EL2, ATP6V1EL2, MGC9341, VMA4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-62363