Learn More
Aviva Systems Biology Corp ATP6V1G2 Antibody - middle region (ARP57723_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57723_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP6V1G2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATP6V1G2. Alternate names: ATP6G, ATP6G2, NG38, VMA10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-286-363