Learn More
Aviva Systems Biology Corp ATP8B2 Antibody - N-terminal region (ARP44521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATP8B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ATP8B2. Alternate names: ATPID, DKFZp434M0219, KIAA1137ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-148