Learn More
Medchemexpress LLC HY-101261 10mg Medchemexpress, ATPA CAS:140158-50-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10126110MG
Encompass_Preferred
Medchemexpress, HY-101261 10mg ATPA CAS:140158-50-5 ATPA is a selective glutamate receptor GluR5 activator with EC50s of 0.66, 9.5, 1.4, 23, 32, 18, and 14 μM for GluR5wt, GluR5(S741M), GluR5(S721T), GluR5(S721T, S741M), GluR5(S741A), GluR5(S741L), and GluR5(S741V), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7819