Learn More
Aviva Systems Biology Corp ATPAF1 Antibody - middle region (ARP57652_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57652_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ATPAF1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ATPAF1. Alternate names: ATP11, ATP11p, FLJ22351, MGC88060ApplicationWB. Species reactivity: Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-343