Learn More
Medchemexpress LLC HY-126653 1mg , Atpenin A5 CAS:119509-24-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1266531MG
Encompass_Preferred
HY-126653 1mg Atpenin A5 CAS: 119509-24-9 Atpenin A5 is a potent and highly specific complex II inhibitor (IC50 ~10 nM), and is an effective mKATP channel agonist and cardioprotective agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1344