Learn More
Eurofins Discoverx ATR/ATRIP, active, 250 µg. (14-953M)
Supplier:  Eurofins Discoverx 14953M
Encompass
Recombinant, human FLAG-tagged ATR full length and recombinant, human cMyc-tagged ATRIP full length. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9475
Add to Cart
Request a Quote