Learn More
Medchemexpress LLC ATR kinase substrate peptide | 1232497-16-3 | 99.3% | 1814.05 | 1 MG
SDP
Catalog No. 5000425141
Encompass_Preferred
ATR kinase substrate peptide (ASELPASQPQPFSAKKK) is a peptide substrate for ATR protein kinase. It can be utilized to detect ATR kinase activity.
  • Functions as a peptide substrate for ATR protein kinase
  • Utilized to detect ATR kinase activity
  • Has a Ki value of 6 nM against ATR
  • Features the sequence shortening ASELPASQPQPFSAKKK
Catalog No. 50-004-25141 Supplier Medchemexpress LLC Supplier No. HYP102801MG
ATR kinase substrate peptide (ASELPASQPQPFSAKKK) is a peptide substrate for ATR protein kinase. It can be utilized to detect ATR kinase activity.
  • Functions as a peptide substrate for ATR protein kinase
  • Utilized to detect ATR kinase activity
  • Has a Ki value of 6 nM against ATR
  • Features the sequence shortening ASELPASQPQPFSAKKK