Learn More
Perkin Elmer US LLC ATR MIRacle Accessory for the Spectrum Two - Single Reflection, ZnSe
Catalog No. 502113128
Encompass_Preferred
The MIRacle ATR accessory is a single reflection HATR accessory that specializes in the analysis of highly absorbing solid, semi-liquid, and liquid samples. The sampling plate's small round crystal ensures reliable analysis of small sample volumes.
Catalog No. 50-211-3128 Supplier Perkin Elmer US LLC Supplier No. L1600115
Add to Cart
Add to Cart
Request a Quote
The MIRacle ATR accessory is a single reflection HATR accessory that specializes in the analysis of highly absorbing solid, semi-liquid, and liquid samples. The sampling plate's small round crystal ensures reliable analysis of small sample volumes.