Learn More
Medchemexpress LLC ATRA-biotin | 2226143-93-5 | 98.4% | C36H55N3O4S | 100 MG
SDP
Catalog No. 5000353426
Encompass_Preferred
ATRA-biotin is a biotin-conjugated ATRA that can be used to track ATRA in cells or a given tissue. It has demonstrated biocompatibility via cell viability assays in cultured N2a-cells.
  • Biotin-conjugated ATRA
  • Enables tracking of ATRA in cells or tissues
  • Biocompatible, as shown by cell viability assays in cultured N2a-cells
Catalog No. 50-003-53426 Supplier Medchemexpress LLC Supplier No. HY141793100MG
ATRA-biotin is a biotin-conjugated ATRA that can be used to track ATRA in cells or a given tissue. It has demonstrated biocompatibility via cell viability assays in cultured N2a-cells.
  • Biotin-conjugated ATRA
  • Enables tracking of ATRA in cells or tissues
  • Biocompatible, as shown by cell viability assays in cultured N2a-cells