Learn More
Medchemexpress LLC ATRA-biotin (biotin-ATRA-conjugate) | 2226143-93-5 | 98.4% | 625.90 | C36H55N3O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1417935MG
Encompass_Preferred
ATRA-biotin is a biotin-conjugated derivative of all-trans retinoic acid designed to track ATRA localization and interactions in cells and tissues. The biotin moiety permits detection and affinity capture using streptavidin/avidin reagents, facilitating imaging, pull-down, and binding studies. The compound is supplied at high reported purity and includes documentation for characterization and safe handling.
  • Biotin tag enables streptavidin-based detection and affinity capture.
  • Suitable for tracking ATRA localization in cells and tissue samples.
  • High reported purity supports biochemical and cell-based assays.
  • Available in multiple pack sizes for scalability of experiments.
  • Provided with datasheet, COA, and SDS for characterization and safety.
Catalog No. 50-002-21507