Learn More
Cayman Chemical AtractylenolIde I 10mg
SDP
Catalog No. 502933964
Encompass_Preferred
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited
Catalog No. 50-293-3964 Supplier Cayman Chemical Supplier No. 2506110 MG
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited