Learn More
Medchemexpress LLC HY-N0201 10mg Medchemexpress, Atractylenolide I CAS:73069-13-3 Purity:>98%
SDP
Catalog No. 501933462
Encompass_Preferred
Medchemexpress, HY-N0201 10mg Atractylenolide I CAS:73069-13-3 Atractylenolide I is a sesquiterpene derived from the rhizome of Atractylodes macrocephala, possesses diverse bioactivities, such as neuroprotective, anti-allergic, anti-inflammatory and anticancer properties. Atractylenolide I reduces protein levels of phosphorylated JAK2 and STAT3 in A375 cells, and acts as a TLR4 -antagonizing agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3462 Supplier Medchemexpress LLC Supplier No. HYN020110MG
Medchemexpress, HY-N0201 10mg Atractylenolide I CAS:73069-13-3 Atractylenolide I is a sesquiterpene derived from the rhizome of Atractylodes macrocephala, possesses diverse bioactivities, such as neuroprotective, anti-allergic, anti-inflammatory and anticancer properties. Atractylenolide I reduces protein levels of phosphorylated JAK2 and STAT3 in A375 cells, and acts as a TLR4 -antagonizing agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.