Learn More
Cayman Chemical AtractylenolIde I 25mg
SDP
Catalog No. 502933953
Encompass_Preferred
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited
Catalog No. 50-293-3953 Supplier Cayman Chemical Supplier No. 2506125 MG
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited