Learn More
Cayman Chemical AtractylenolIde I 50mg
SDP
Catalog No. 502934852
Encompass_Preferred
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited
Catalog No. 50-293-4852 Supplier Cayman Chemical Supplier No. 2506150 MG
An acaricide effective against populations of R. microplus R. sanguineus S. scabei and Y. pestis at host doses ranging from 1.5 to 100 mg/kg adverse ecotoxicological effect may be limited