Learn More
Cayman Chemical Atractylodin, Purity: ≥98%, CAS: 55290-63-6, 50 mg
SDP
Catalog No. 502913133
Encompass_Preferred
A polyethylene alkyne with diverse biological activities; inhibits NAAA (IC50 = 2.81 µM for the human enzyme); increases levels of palmitoyl ethanolamide and oleoyl ethanolamide in LPS-stimulated BV-2 cells at 10 µM; cytotoxic to CL-6 and HuCC-T1 cells (IC50s = 41.66 and 38.78 µM, respectively); increases the gastric emptying rate in mice at 10 mg/kg; decreases pulmonary edema in a mouse model of LPS-induced acute lung injury at 40 and 80 mg/kg
Catalog No. 50-291-3133 Supplier Cayman Chemical Supplier No. 3503350 MG
A polyethylene alkyne with diverse biological activities; inhibits NAAA (IC50 = 2.81 µM for the human enzyme); increases levels of palmitoyl ethanolamide and oleoyl ethanolamide in LPS-stimulated BV-2 cells at 10 µM; cytotoxic to CL-6 and HuCC-T1 cells (IC50s = 41.66 and 38.78 µM, respectively); increases the gastric emptying rate in mice at 10 mg/kg; decreases pulmonary edema in a mouse model of LPS-induced acute lung injury at 40 and 80 mg/kg