Learn More
Medchemexpress LLC HY-N1462 1mg Medchemexpress, Atractyloside (potassium salt) CAS:102130-43-8 Purity:>98%
SDP
Catalog No. 501970343
Encompass_Preferred
Medchemexpress, HY-N1462 1mg Atractyloside (potassium salt) CAS:102130-43-8 Atractyloside potassium salt is a toxic diterpenoid glycoside isolated from the fruits of Xanthium sibiricum. Atractyloside potassium salt is a powerful and specific inhibitor of mitochondrial ADP/ATP transport. Atractyloside potassium salt inhibits chloride channels from mitochondrial membranes of rat heart[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0343 Supplier Medchemexpress LLC Supplier No. HYN14621MG
Medchemexpress, HY-N1462 1mg Atractyloside (potassium salt) CAS:102130-43-8 Atractyloside potassium salt is a toxic diterpenoid glycoside isolated from the fruits of Xanthium sibiricum. Atractyloside potassium salt is a powerful and specific inhibitor of mitochondrial ADP/ATP transport. Atractyloside potassium salt inhibits chloride channels from mitochondrial membranes of rat heart[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.