Learn More
Selleck Chemical LLC Atractyloside potassium salt S3264-1mg
SDP
Catalog No. 503110439
Encompass_Preferred
Atractyloside potassium salt (ATR potassium salt) a toxic diterpenoid glycoside isolated from the fruits of Xanthium sibiricum (Cang erzi) is a powerful and specific inhibitor of mitochondrial ADP/ATP carriers Atractyloside potassium salt inhibits chloride channels from mitochondrial membranes of rat heart
Catalog No. 50-311-0439 Supplier Selleck Chemical LLC Supplier No. S32641MG
Atractyloside potassium salt (ATR potassium salt) a toxic diterpenoid glycoside isolated from the fruits of Xanthium sibiricum (Cang erzi) is a powerful and specific inhibitor of mitochondrial ADP/ATP carriers Atractyloside potassium salt inhibits chloride channels from mitochondrial membranes of rat heart