Learn More
TARGETMOL CHEMICALS INC Atracurium besylate 100MG
SDP
Catalog No. 502879838
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Atracurium besylate (51W89) is a non-depolarizing neuromuscular blocking agent with short duration of action. Its lack of significant cardiovascular effects and its lack of dependence on good kidney function for elimination provide clinical advantage over alternate non-depolarizing neuromuscular blocking agents. Purity 99.71%
Catalog No. 50-287-9838 Supplier TARGETMOL CHEMICALS INC Supplier No. T6397100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Atracurium besylate (51W89) is a non-depolarizing neuromuscular blocking agent with short duration of action. Its lack of significant cardiovascular effects and its lack of dependence on good kidney function for elimination provide clinical advantage over alternate non-depolarizing neuromuscular blocking agents. Purity 99.71%