Learn More
Selleck Chemical LLC Atracurium Besylate S1832-50mg
SDP
Catalog No. 503110822
Encompass_Preferred
Atracurium Besylate (BW 33A 51W89) is a neuromuscular blocking agent with ED95 of 0 2 mg/kg
Catalog No. 50-311-0822 Supplier Selleck Chemical LLC Supplier No. S183250MG
Atracurium Besylate (BW 33A 51W89) is a neuromuscular blocking agent with ED95 of 0 2 mg/kg