Learn More
Apexbio Technology LLC Atracurium Besylate 64228-81-5 500mg
SDP
Catalog No. 5000088270
Encompass_Preferred
Atracurium Besylate (CAS 64228-81-5) is a non-depolarizing neuromuscular blocking agent that exerts its effects by competitively inhibiting acetylcholine receptors at the neuromuscular junction thereby interrupting neurotransmission between motor nerves and skeletal muscle In biomedical research it is utilized to induce neuromuscular blockade enabling studies of neurotransmitter release synaptic transmission mechanisms and regulation of muscle contraction Additionally Atracurium Besylate has been reported to elicit bronchoconstriction and promote differentiation of glial cells such as astrocytes supporting investigations into cellular differentiation and airway responsiveness
Catalog No. 50-000-88270 Supplier Apexbio Technology LLC Supplier No. B1666500
Atracurium Besylate (CAS 64228-81-5) is a non-depolarizing neuromuscular blocking agent that exerts its effects by competitively inhibiting acetylcholine receptors at the neuromuscular junction thereby interrupting neurotransmission between motor nerves and skeletal muscle In biomedical research it is utilized to induce neuromuscular blockade enabling studies of neurotransmitter release synaptic transmission mechanisms and regulation of muscle contraction Additionally Atracurium Besylate has been reported to elicit bronchoconstriction and promote differentiation of glial cells such as astrocytes supporting investigations into cellular differentiation and airway responsiveness