Learn More
Cayman Chemical Atracurium (besylate), Purity: ≥95%, CAS: 64228-81-5, 10 mg
SDP
Catalog No. 502967591
Encompass_Preferred
A benzylisoquinoline that acts as a neuromuscular blocking agent with slow onset and intermediate duration; causes non-depolarizing block of the neuromuscular junction
Catalog No. 50-296-7591 Supplier Cayman Chemical Supplier No. 1779610 MG
A benzylisoquinoline that acts as a neuromuscular blocking agent with slow onset and intermediate duration; causes non-depolarizing block of the neuromuscular junction