Learn More
Cayman Chemical AtracurIumbesylate 50mg
SDP
Catalog No. 502966419
Encompass_Preferred
A benzylisoquinoline that acts as a neuromuscular blocking agent with slow onset and intermediate duration; causes non-depolarizing block of the neuromuscular junction
Catalog No. 50-296-6419 Supplier Cayman Chemical Supplier No. 1779650 MG
A benzylisoquinoline that acts as a neuromuscular blocking agent with slow onset and intermediate duration; causes non-depolarizing block of the neuromuscular junction