Learn More
Medchemexpress LLC Atrasentan | 173937-91-2 | C29H38N2O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1540310MM/1ML
Encompass_Preferred
Atrasentan is an endothelin receptor antagonist known for its potent activity against the ETA receptor, with an IC50 of 0.0551 nM. Also identified as ABT-627, it plays a role in the GPCR/G Protein pathway and has a molecular formula of C29H38N2O6 and a molecular weight of 510.62. This compound is utilized in research studying various biological activities.
  • Significantly inhibits LNCaP and C4-2b prostate cancer cell growth
  • Enhances down-regulation of NF-κB DNA binding activity when combined with Taxotere
  • Induces several CYPs and drug transporters, such as CYP3A4
  • Functions as a moderate P-gp inhibitor and a weak BCRP inhibitor
  • Inhibits pressor response induced by big endothelin-1 in pithed rats
  • Inhibits C4-2b tumor growth within the bone environment in the SCID-hu model
Catalog No. 50-003-76328