Learn More
Medchemexpress LLC Atrasentan | 173937-91-2 | C29H38N2O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY15403
Encompass_Preferred
Atrasentan (ABT-627) is an endothelin receptor antagonist with an IC50 of 0.0551 nM for ETA. It significantly inhibits LNCaP and C4-2b prostate cancer cell growth in vitro and demonstrates anti-tumor effects in vivo. It also induces several CYPs and acts as a moderate P-gp inhibitor.
  • Endothelin A receptor antagonist with an IC50 of 0.0551 nM for ETA
  • Inhibits LNCaP and C4-2b prostate cancer cell growth
  • Elicits greater loss of viable prostate cancer cells when combined with Taxotere
  • Down-regulates NF-κB DNA binding activity
  • Induces several CYPs and drug transporters (e.g., CYP3A4)
  • Moderate P-gp inhibitor (IC50 = 15.1 ± 1.6 μM in P388/dx cells)
  • Weak BCRP inhibitor (IC50 = 59.8 ± 11 μM in MDCKII-BCRP cells)
  • Inhibits pressor response induced by big endothelin-1 in pithed rats
  • Inhibits C4-2b tumor growth within the bone environment in the SCID-hu model
Catalog No. 50-003-74897