Learn More
Medchemexpress LLC Atrasentan | 173937-91-2 | C29H38N2O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY154035MG
Encompass_Preferred
Atrasentan (ABT-627) is an endothelin receptor antagonist that targets ETA with an IC50 of 0.055 nM. It significantly inhibits LNCaP and C4-2b prostate cancer cell growth and also induces several CYPs and drug transporters. It is intended for research use only.
  • Endothelin receptor antagonist
  • Targets ETA receptor
  • Inhibits LNCaP and C4-2b prostate cancer cell growth
  • Down-regulates NF-κB DNA binding activity
  • Induces CYPs and drug transporters
  • Moderate P-gp inhibitor
  • Weak BCRP inhibitor
  • Inhibits pressor response induced by big endothelin-1 in pithed rats
  • Inhibits C4-2b tumor growth within the bone environment
Catalog No. 50-003-76349