Learn More
Medchemexpress LLC Atrasentan hydrochloride | 195733-43-8 | C29H39ClN2O6 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY15403A10MM/1ML
Encompass_Preferred
Atrasentan hydrochloride is a selective endothelin A receptor antagonist, primarily used for research purposes. It has demonstrated significant inhibition of LNCaP and C4-2b prostate cancer cell growth in vitro, with an IC50 of 0.0551 nM for ETA. This compound also impacts drug metabolism by inducing several CYPs and drug transporters.
  • Selectively antagonizes endothelin A receptors
  • Inhibits LNCaP and C4-2b prostate cancer cell growth
  • Induces CYPs and drug transporters
  • Acts as a moderate P-gp inhibitor and weak BCRP inhibitor
  • Demonstrates in vivo inhibition of pressor response induced by big endothelin-1
  • Inhibits C4-2b tumor growth in SCID-hu model
Catalog No. 50-003-76174