Learn More
Apexbio Technology LLC ATRASENTAN-HYDROCHLORIDE-2MG
SDP
Catalog No. 5000568003
Encompass_Preferred
Atrasentan hydrochloride (CAS No 195733-43-8) is a potent compound used in biomedical research particularly within the field of oncology and nephrology It is an endothelin receptor antagonist primarily targeting the ETA receptors involved in vasoconstriction and cell proliferation pathways This compound exhibits activity in the nanomolar to low micromolar range which makes it suitable for precise in vitro experiments Atrasentan hydrochloride is commonly utilized in a variety of applications including cell model studies animal research and drug screening processes Its role in inhibiting endothelin-1 mediated pathophysiological processes makes it valuable for studying diseases such as cancer and chronic kidney disease Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse experimental setups
Catalog No. 50-005-68003 Supplier Apexbio Technology LLC Supplier No. A32012
Atrasentan hydrochloride (CAS No 195733-43-8) is a potent compound used in biomedical research particularly within the field of oncology and nephrology It is an endothelin receptor antagonist primarily targeting the ETA receptors involved in vasoconstriction and cell proliferation pathways This compound exhibits activity in the nanomolar to low micromolar range which makes it suitable for precise in vitro experiments Atrasentan hydrochloride is commonly utilized in a variety of applications including cell model studies animal research and drug screening processes Its role in inhibiting endothelin-1 mediated pathophysiological processes makes it valuable for studying diseases such as cancer and chronic kidney disease Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse experimental setups