Learn More
Sklar Instruments Debakey Atraumatic Forceps, Sklar 14 in., 2.7mm, Blunt, Straight, 1X2 Rows of Nontraumatic Fine Teeth. EA.
SDP
Catalog No. 501181432 Shop All Sklar Instruments Products
Encompass_Preferred
Debakey Atraumatic Forceps, Sklar. 14 in., 2.7mm, Blunt, Straight, 1X2 Rows of Nontraumatic Fine Teeth. Sklar DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used in numerous surgical procedures. They are commonly used for grasping fine tissue and blood vessels or soft tissue dissection. The narrow tips and atraumatic teeth make these forceps particularly useful in cardiothoracic, vascular, laparotomy, and plastic surgery procedures where it is necessary to handle delicate tissues. DeBakey Atraumatic Tissue Forceps are available in multiple lengths, straight or angled, and with a variety of tip dimensions. This is one more reason why they are a popular and useful surgical instrument. This product is straight with 1x2 rows of fine, atraumatic teeth on 2.7mm jaws. The overall length is 14 inches. They are frequently used in bariatric procedures.
Catalog No. 50-118-1432 Supplier Sklar Instruments Supplier No. 525184
Debakey Atraumatic Forceps, Sklar. 14 in., 2.7mm, Blunt, Straight, 1X2 Rows of Nontraumatic Fine Teeth. Sklar DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used in numerous surgical procedures. They are commonly used for grasping fine tissue and blood vessels or soft tissue dissection. The narrow tips and atraumatic teeth make these forceps particularly useful in cardiothoracic, vascular, laparotomy, and plastic surgery procedures where it is necessary to handle delicate tissues. DeBakey Atraumatic Tissue Forceps are available in multiple lengths, straight or angled, and with a variety of tip dimensions. This is one more reason why they are a popular and useful surgical instrument. This product is straight with 1x2 rows of fine, atraumatic teeth on 2.7mm jaws. The overall length is 14 inches. They are frequently used in bariatric procedures.