Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atrazine | 1912-24-9 | 500 MG
SDP

Catalog No. 5000420971
Encompass_Preferred

Atrazine is a triazine herbicide primarily used to control certain annual broadleaf and grass weeds. It functions by inhibiting photophosphorylation but typically does not cause lethality or permanent cell damage in the short term.

  • Controls certain annual broadleaf and grass weeds
  • Inhibits photophosphorylation
  • Purity of 99.32%
  • Molecular weight of 215.68
  • Molecular formula C8H14ClN5
  • Appears as a white to off-white solid
  • Store solid at 4°C, sealed and away from moisture
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Catalog No. 50-004-20971 Supplier Medchemexpress LLC Supplier No. HYN7091500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Atrazine is a triazine herbicide primarily used to control certain annual broadleaf and grass weeds. It functions by inhibiting photophosphorylation but typically does not cause lethality or permanent cell damage in the short term.

  • Controls certain annual broadleaf and grass weeds
  • Inhibits photophosphorylation
  • Purity of 99.32%
  • Molecular weight of 215.68
  • Molecular formula C8H14ClN5
  • Appears as a white to off-white solid
  • Store solid at 4°C, sealed and away from moisture
  • For solvent solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.