Learn More
Chem Service Inc Atrazine,1912-24-9,250MG
SDP
Catalog No. NC0051573
Encompass_Preferred
Molecular Formula C8H14ClN5; Purity 99.10%; Molecular Weight 215.69; Melting Point 175 - 177 °C; Synonyms: 6-Chloro-N-ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diaminel;
Catalog No. NC0051573 Supplier Chem Service Inc Supplier No. N11106250MG
Molecular Formula C8H14ClN5; Purity 99.10%; Molecular Weight 215.69; Melting Point 175 - 177 °C; Synonyms: 6-Chloro-N-ethyl-N'-(1-methylethyl)-1,3,5-triazine-2,4-diaminel;