Learn More
Research Products International Corp Atrazine, 250 Milligrams
SDP
Catalog No. 50213158
Encompass_Preferred
Inhibits photosynthesis by blocking the electron transport. Binds to Qb protein in the thyalkoid membrane.
Catalog No. 50-213-158 Supplier Research Products International Corp Supplier No. A600200.25
Inhibits photosynthesis by blocking the electron transport. Binds to Qb protein in the thyalkoid membrane.