Learn More
Medchemexpress LLC Atrazine (Standard) | 1912-24-9 | 100 MG
SDP
Catalog No. 5000418864
Encompass_Preferred
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications, such as controlling certain annual broadleaf and grass weeds. It functions by inhibiting photophosphorylation and serves as a reference standard for various assays.
  • Intended for research and analytical applications
  • Controls annual broadleaf and grass weeds
  • Inhibits photophosphorylation
  • Reference standard for assays
  • Commonly used in HPLC, GC, and MS experiments
Catalog No. 50-004-18864 Supplier Medchemexpress LLC Supplier No. HYN7091R100MG
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications, such as controlling certain annual broadleaf and grass weeds. It functions by inhibiting photophosphorylation and serves as a reference standard for various assays.
  • Intended for research and analytical applications
  • Controls annual broadleaf and grass weeds
  • Inhibits photophosphorylation
  • Reference standard for assays
  • Commonly used in HPLC, GC, and MS experiments