Learn More
Medchemexpress LLC Atrazine (Standard) | 1912-24-9 | 250 MG
SDP
Catalog No. 5000422084
Encompass_Preferred
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications. This compound is known for controlling certain annual broadleaf and grass weeds by inhibiting photophosphorylation, without typically causing lethality or permanent cell damage in the short term.
  • Intended for research and analytical applications
  • Analytical standard grade
  • Used as a reference standard in assays
  • Commonly applied in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Inhibits photophosphorylation
Catalog No. 50-004-22084 Supplier Medchemexpress LLC Supplier No. 250MGHYN7091R
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications. This compound is known for controlling certain annual broadleaf and grass weeds by inhibiting photophosphorylation, without typically causing lethality or permanent cell damage in the short term.
  • Intended for research and analytical applications
  • Analytical standard grade
  • Used as a reference standard in assays
  • Commonly applied in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Inhibits photophosphorylation