Learn More
Medchemexpress LLC Atrazine (Standard) | 1912-24-9 | 50 MG
SDP
Catalog No. 5000419685
Encompass_Preferred
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications. This compound is effective in controlling certain annual broadleaf and grass weeds by inhibiting photophosphorylation. It serves as an analytical grade standard, crucial for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Primarily used for control of certain annual broadleaf and grass weeds
  • Functions by inhibiting photophosphorylation
  • Serves as an analytical grade standard for HPLC, GC, and MS
Catalog No. 50-004-19685 Supplier Medchemexpress LLC Supplier No. HYN7091R50MG
Atrazine (Standard) is an analytical standard primarily used for research and analytical applications. This compound is effective in controlling certain annual broadleaf and grass weeds by inhibiting photophosphorylation. It serves as an analytical grade standard, crucial for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Primarily used for control of certain annual broadleaf and grass weeds
  • Functions by inhibiting photophosphorylation
  • Serves as an analytical grade standard for HPLC, GC, and MS