Learn More
Cayman Chemical Atrazn-d5 10mg
SDP
Catalog No. 502916202
Encompass_Preferred
An internal standard for the quantification of atrazine by GC- or LC-MS
Catalog No. 50-291-6202 Supplier Cayman Chemical Supplier No. 3955110 MG
An internal standard for the quantification of atrazine by GC- or LC-MS