Learn More
Medchemexpress LLC HY-P1236 1mg Medchemexpress, Atrial Natriuretic Peptide (ANP) (1-28), rat CAS:88898-17-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP12361MG
Encompass_Preferred
Medchemexpress, HY-P1236 1mg Atrial Natriuretic Peptide (ANP) (1-28), rat CAS:88898-17-3 Atrial Natriuretic Peptide (ANP) (1-28), rat is a major circulating form of ANP in rats, potently inhibits Angiotensin II (Ang II)-stimulated endothelin-1 secretion in a concentration-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4175