Learn More
Medchemexpress LLC HY-P1235A 1mg Medchemexpress, Atrial Natriuretic Peptide (ANP) (1-28), human, porcine (Acetate) CAS:1366000-58-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP1235A1MG
Encompass_Preferred
Medchemexpress, HY-P1235A 1mg Atrial Natriuretic Peptide (ANP) (1-28), human, porcine (Acetate) CAS:1366000-58-9 Atrial Natriuretic Peptide (ANP) (1-28), human, porcine Acetate is a 28-amino acid hormone, that is normally produced and secreted by the human heart in response to cardiac injury and mechanical stretch. ANP (1-28) inhibits endothelin-1 secretion in a dose-dependent way. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4174