Learn More
CPC Scientific H-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Ile-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR003B
Encompass
ONE-LETTER SEQUENCE: SLRRSSCFGGRIDRIGAQSGLGCNSFRY (Cys7 and 23 bridge)MOLECULAR FORMULA: C128H205N45O39S2MOLECULAR WEIGHT:3062.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [88898-17-3], RESEARCH AREA: CardiovascularREFERENCES: T.G. Flynn, et al., BBRC, 117, 859 (1983)
Catalog No. 50-210-9852