Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Atrial natriuretic peptide (1-28), human, porcine, biotin-labeled | 1815618-06-4 | 99.4% | 3308.80 | 1 MG
SDP

Catalog No. 5000423964
Encompass_Preferred

Atrial natriuretic peptide (1-28), human, porcine, biotin-labeled, is one of three mammalian natriuretic peptides (NPs) and has endocrine effects on fluid homeostasis and blood pressure. It has potential for cardiovascular diseases research.

  • Purity: 99.43%
  • Molecular weight: 3308.80
  • Formula: C137H217N47O41S4
  • CAS no.: 1815618-06-4
  • Appearance: Solid, white to off-white
  • Sequence: Biotin-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr (disulfide bridge: Cys7-Cys23)
  • Sequence shortening: Biotin-SLRRSSCFGGRMDRIGAQSGLGCNSFRY (disulfide bridge: Cys7-Cys23)
  • Solubility (in vitro): H2O: 5 mg/mL (1.51 mM; needs ultrasonic)

Catalog No. 50-004-23964 Supplier Medchemexpress LLC Supplier No. HYP24911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Atrial natriuretic peptide (1-28), human, porcine, biotin-labeled, is one of three mammalian natriuretic peptides (NPs) and has endocrine effects on fluid homeostasis and blood pressure. It has potential for cardiovascular diseases research.

  • Purity: 99.43%
  • Molecular weight: 3308.80
  • Formula: C137H217N47O41S4
  • CAS no.: 1815618-06-4
  • Appearance: Solid, white to off-white
  • Sequence: Biotin-Ser-Leu-Arg-Arg-Ser-Ser-Cys-Phe-Gly-Gly-Arg-Met-Asp-Arg-Ile-Gly-Ala-Gln-Ser-Gly-Leu-Gly-Cys-Asn-Ser-Phe-Arg-Tyr (disulfide bridge: Cys7-Cys23)
  • Sequence shortening: Biotin-SLRRSSCFGGRMDRIGAQSGLGCNSFRY (disulfide bridge: Cys7-Cys23)
  • Solubility (in vitro): H2O: 5 mg/mL (1.51 mM; needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.