Learn More
Medchemexpress LLC Atrial natriuretic peptide (1-28), human, porcine, biotin-labeled | 1815618-06-4 | 99.4% | 3308.80 | 5 MG
SDP
Catalog No. 5000423578
Encompass_Preferred
Atrial Natriuretic Peptide (1-28), human, porcine, Biotin-labeled is one of three mammalian natriuretic peptides (NPs) that exerts endocrine effects on fluid homeostasis and blood pressure. This product holds potential for cardiovascular diseases research.
  • Exerts endocrine effects on fluid homeostasis and blood pressure
  • Potential for cardiovascular diseases research
  • High purity of 99.43%
  • White to off-white solid appearance
  • Biotin-labeled sequence with a disulfide bridge (Cys7-Cys23)
Catalog No. 50-004-23578 Supplier Medchemexpress LLC Supplier No. HYP24915MG
Atrial Natriuretic Peptide (1-28), human, porcine, Biotin-labeled is one of three mammalian natriuretic peptides (NPs) that exerts endocrine effects on fluid homeostasis and blood pressure. This product holds potential for cardiovascular diseases research.
  • Exerts endocrine effects on fluid homeostasis and blood pressure
  • Potential for cardiovascular diseases research
  • High purity of 99.43%
  • White to off-white solid appearance
  • Biotin-labeled sequence with a disulfide bridge (Cys7-Cys23)