Learn More
Genscript Corporation Atrial Natriuretic Peptide (ANP) (1-28), human, porcine
Supplier:  Genscript Corporation RP119270.5
Encompass
Atrial natriuretic peptide (ANP), a stimulator of particulate guanylate cyclase, has been found to protect against reoxygenation-induced hypercontracture in isolated cardiomyocytes by increasing cyclic guanosine monophosphate synthesis; ANP also contributes to stimulation of lipolysis during repeated bouts of exercise and in the control of lipolysis during exercise.
Catalog No. 50-275-155