Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Atrial Natriuretic Peptide, Fragment 3-28, 98.1%, MP Biomedicals

Catalog No. ICN15989080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15989080 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15989080 Supplier MP Biomedicals, Inc Supplier No. 0215989080
Add to Cart
Add to Cart

Specifications

Assay 0.981
CAS 90984-99-9
Color White
Format Lyophilized Powder
Molecular Weight (g/mol) 2862.206
Product Type Atrial Natriuretic Peptide, Fragment 3-28
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.