Learn More
ECM Bioscience Atrogin-1 Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AP2041
Encompass_Preferred
This antibody detects a 41 kDa* protein corresponding to the apparent molecular mass of Atrogin on SDS-PAGE immunoblots of mouse muscle tissue lysates. Similar results were seen in C2C12 cell lysate and the band observed in western blot is specifically blocked by Atrogin-1 peptide (AX2045). The immunogen was A synthetic peptide (coupled to carrier protein) corresponding to amino acids 23 to 35 in mouse Atrogin. This sequence is highly homologous to human and rat Atrogin (F-box only protein 32). Accession #Q9CPU7. Applications = WB, E, IHC; Species Reactivity = Hu, Rt, Ms; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = MAFbx, FBX32, Atrogin
Catalog No. 50-172-0714