Learn More
Cayman Chemical AtropIn-d5 2.5mg
SDP
Catalog No. 502989644
Encompass_Preferred
An internal standard for the quantification of atropine by GC- or LC-MS
Catalog No. 50-298-9644 Supplier Cayman Chemical Supplier No. 307342.5 MG
An internal standard for the quantification of atropine by GC- or LC-MS